Learn More
Ted Pella Inc AFM/STM Cantilever Tweezers, Titanium
SDP
Catalog No. NC1262074
Encompass
Easily grasp magnetic AFM/STM cantilevers / probes with these precise titanium, absolutely non-magnetic tweezers. 4-5/8" (117mm) long.
Catalog No. NC1262074 Supplier Ted Pella Inc Supplier No. 5596TI
Easily grasp magnetic AFM/STM cantilevers / probes with these precise titanium, absolutely non-magnetic tweezers. 4-5/8" (117mm) long.