Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Afm32a hydrochloride | 2988594-85-8 | 99.4% | 517.00 | 1 MG
SDP

Catalog No. 5000351908
Encompass_Preferred

Afm32a hydrochloride | 2988594-85-8 | 99.4% | 517.00 | 1 MG

Catalog No. 50-003-51908 Supplier Medchemexpress LLC Supplier No. HY136557A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Afm32a hydrochloride | 2988594-85-8 | 99.4% | 517.00 | 1 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.