Learn More
Medchemexpress LLC AFM32a hydrochloride | 2988594-85-8 | 99.4% | 517.00 | 1 ML
SDP
Catalog No. 5000349403
Encompass_Preferred
AFM32a hydrochloride, also known as PAD2-IN-1 hydrochloride, is a potent and selective benzimidazole-based derivative that inhibits protein arginine deiminase 2 (PAD2). It demonstrates high selectivity, being 95-fold more selective for PAD2 over PAD4 and 79-fold more selective over PAD3.
  • Potent and selective PAD2 inhibitor.
  • Exhibits superior selectivity over PAD4 and PAD3.
  • Effective in inhibiting histone H3 citrullination in HEK293T/PAD2 cells.
Catalog No. 50-003-49403 Supplier Medchemexpress LLC Supplier No. HY136557A10MM/1ML
AFM32a hydrochloride, also known as PAD2-IN-1 hydrochloride, is a potent and selective benzimidazole-based derivative that inhibits protein arginine deiminase 2 (PAD2). It demonstrates high selectivity, being 95-fold more selective for PAD2 over PAD4 and 79-fold more selective over PAD3.
  • Potent and selective PAD2 inhibitor.
  • Exhibits superior selectivity over PAD4 and PAD3.
  • Effective in inhibiting histone H3 citrullination in HEK293T/PAD2 cells.