Learn More
Medchemexpress LLC Afm32a hydrochloride | 2988594-85-8 | 99.7% | 517.00 g/mol | C25H30ClFN6O3 | 5 MG
SDP
Catalog No. 5000222273
Encompass_Preferred
AFM32a hydrochloride is a benzimidazole-derived small-molecule inhibitor selective for protein arginine deiminase 2 (PAD2). Supplied as the hydrochloride salt for research use, it is suitable for biochemical assays and target validation, demonstrating approximately 95-fold selectivity for PAD2 versus PAD4 and 79-fold versus PAD3.
  • Potent PAD2 inhibition with high selectivity over PAD3 and PAD4.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity (≈99.7%) appropriate for biochemical assays.
  • Available in small-mass and DMSO solution formats for screening.
  • Characterized with molecular formula and molecular weight for analytical use.
Catalog No. 50-002-22273 Supplier Medchemexpress LLC Supplier No. HY136557A5MG
AFM32a hydrochloride is a benzimidazole-derived small-molecule inhibitor selective for protein arginine deiminase 2 (PAD2). Supplied as the hydrochloride salt for research use, it is suitable for biochemical assays and target validation, demonstrating approximately 95-fold selectivity for PAD2 versus PAD4 and 79-fold versus PAD3.
  • Potent PAD2 inhibition with high selectivity over PAD3 and PAD4.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity (≈99.7%) appropriate for biochemical assays.
  • Available in small-mass and DMSO solution formats for screening.
  • Characterized with molecular formula and molecular weight for analytical use.