Learn More
Cayman Chemical AFP 07free acId 500ug
SDP
Catalog No. 502955141
Encompass_Preferred
A 7,7-difluoroprostacyclin derivative that acts as a selective and highly potent agonist for the IP receptor (Ki = 0.561 nM); shows weaker affinity for EP receptors, with Ki values of > 100 nM for EP1-3 and > 10 nM for EP4
Catalog No. 50-295-5141 Supplier Cayman Chemical Supplier No. 13626500 UG
A 7,7-difluoroprostacyclin derivative that acts as a selective and highly potent agonist for the IP receptor (Ki = 0.561 nM); shows weaker affinity for EP receptors, with Ki values of > 100 nM for EP1-3 and > 10 nM for EP4