Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 174MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C3. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Hep-G2 cells. Fetal liver or hepatocellular carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Alpha fetoprotein (AFP) purified from serum of a hepatoma patient. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 70kDa.Chromosome: 4q13.3. Gene Symbol: AFP. Gene ID: 174. Ensemble: ENSG00000081051. Uniprot: NP_001341646.
Catalog No. 50-188-4638