Learn More
Sino Biological AFP / alpha-fetoprotein Antibody (APC), Mouse MAb 100Tests
Supplier:  Sino Biological 12177MM35TA100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells. Then conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-224-7751