Learn More
Sino Biological AFP / alpha-fetoprotein Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12177T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human AFP / alpha-fetoprotein (rh AFP / alpha-fetoprotein; Catalog#12177-H08H; P02771; Met1-Val609). AFP / alpha-fetoprotein specific IgG was purified by Human AFP / alpha-fetoprotein affinity chromatography.
Catalog No. 50-224-8178