Learn More
Enzo Life Sciences alpha-Fetoprotein Recombinant monoclonal antibody (AF5) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6080200
Encompass_Preferred
Clone: AF5. Isotype: IgGκ. Immunogen: AF5 was prepared by immunizing female BALB/c mice with human amniotic fluid containing AFP, from a pregnancy affected by a neural tube defect, diluted and emulsified in Freund's complete adjuvant. Species reactivity: Human. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3134