Learn More
Trajan Scientific AFS Purifier I non-hazardous replacement cartridge (not for air or O2 use)
SDP
Catalog No. 502724328
Encompass_Preferred
AFS Purifier I non-hazardous replacement cartridge (not for air or O2 use)
Catalog No. 50-272-4328 Supplier Trajan Scientific Supplier No. 202914
AFS Purifier I non-hazardous replacement cartridge (not for air or O2 use)