Learn More
New England Biolabs, Inc. Afu Uracil-DNA Glycosylase (UDG) – 200 units
SDP
Catalog No. 50995216
Encompass
A thermostable homolog of the E. coli Uracil-DNA Glycosylase (UDG) from Archaeoglobus fulgidus. Afu UDG catalyzes the release of free uracil from uracil-containing DNA. Afu UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA.
Catalog No. 50-995-216 Supplier New England Biolabs, Inc. Supplier No. M0279S
A thermostable homolog of the E. coli Uracil-DNA Glycosylase (UDG) from Archaeoglobus fulgidus. Afu UDG catalyzes the release of free uracil from uracil-containing DNA. Afu UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA.