Learn More
Apexbio Technology LLC Afuresertib 1047644-62-1 100mg
SDP
Catalog No. 5000088690
Encompass_Preferred
Afuresertib (CAS 1047644-62-1) is a small molecule inhibitor targeting the serine/threonine protein kinase Akt (also known as protein kinase B) a key component of the PI3K/Akt signaling pathway involved in cell proliferation survival and metabolism By inhibiting Akt activity afuresertib modulates downstream signaling cascades that contribute to oncogenesis and tumor progression This compound has been investigated in preclinical models and clinical trials for its potential to suppress malignant cell growth particularly in studies involving solid tumors and hematologic malignancies Its use facilitates exploration of Akt pathway inhibition in cancer research
Catalog No. 50-000-88690 Supplier Apexbio Technology LLC Supplier No. B4811100
Afuresertib (CAS 1047644-62-1) is a small molecule inhibitor targeting the serine/threonine protein kinase Akt (also known as protein kinase B) a key component of the PI3K/Akt signaling pathway involved in cell proliferation survival and metabolism By inhibiting Akt activity afuresertib modulates downstream signaling cascades that contribute to oncogenesis and tumor progression This compound has been investigated in preclinical models and clinical trials for its potential to suppress malignant cell growth particularly in studies involving solid tumors and hematologic malignancies Its use facilitates exploration of Akt pathway inhibition in cancer research