Learn More
Selleck Chemical LLC Afuresertib (GSK2110183) 5mg 1047644-62-1
SDP
Supplier:  Selleck Chemical LLC S75215MG
Encompass_Preferred
Afuresertib (GSK2110183) is a potent, orally bioavailable Akt inhibitor with Ki of 0.08 nM, 2 nM, and 2.6 nM for Akt1, Akt2, and Akt3, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0988098