Learn More
Medchemexpress LLC HY-15727A 5mg Medchemexpress, Afuresertib (hydrochloride) CAS:1047645-82-8 Purity:>98%
SDP
Catalog No. 502081779
Encompass_Preferred
Medchemexpress, HY-15727A 5mg Afuresertib (hydrochloride) CAS:1047645-82-8 Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1779 Supplier Medchemexpress LLC Supplier No. HY15727A5MG
Medchemexpress, HY-15727A 5mg Afuresertib (hydrochloride) CAS:1047645-82-8 Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.