Learn More
Medchemexpress LLC HY-N2821 5mg Medchemexpress, (+)-Afzelechin CAS:2545-00-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN28215MG
Encompass_Preferred
Medchemexpress, HY-N2821 5mg (+)-Afzelechin CAS:2545-00-8 (+)-Afzelechin, isolated from rhizomes of Bergenia ligulata, is an alpha-glucosidase activity inhibitor with an ID50 (50% inhibition dose) value of 0.13 mM. (+)-Afzelechin can delay the absorption of carbohydrates in food to suppress postprandial hyperglycemia and hyperinsulinemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2910