Learn More
Medchemexpress LLC Afzelin (Kaempferol-3-O-rhamnoside) | 482-39-3 | MFCD00210589 | 432.38 | 5 MG
SDP
Catalog No. 5000415308
Encompass_Preferred
Afzelin (Kaempferol-3-O-rhamnoside) is a flavonol glycoside with anti-inflammatory, anti-oxidative stress response, anti-apoptotic, and anti-cardiac cytotoxic effects. It can reduce mitochondrial damage, enhance mitochondrial biosynthesis, and mitigate various conditions such as D-galactosamine/LPS induced mortality, doxorubicin-induced cardiotoxicity, scopolamine-induced neurological injury, and ovalbumin-triggered asthma and allergies.
  • Exhibits anti-inflammatory properties.
  • Provides anti-oxidative stress response.
  • Demonstrates anti-apoptotic effects.
  • Offers anti-cardiac cytotoxic effects.
  • Reduces mitochondrial damage.
  • Enhances mitochondrial biosynthesis.
  • Mitigates D-galactosamine/LPS induced mortality.
  • Prevents doxorubicin-induced cardiotoxicity.
  • Ameliorates scopolamine-induced neurological injury.
  • Inhibits asthma and allergies.
Catalog No. 50-004-15308 Supplier Medchemexpress LLC Supplier No. HYN14415MG
Afzelin (Kaempferol-3-O-rhamnoside) is a flavonol glycoside with anti-inflammatory, anti-oxidative stress response, anti-apoptotic, and anti-cardiac cytotoxic effects. It can reduce mitochondrial damage, enhance mitochondrial biosynthesis, and mitigate various conditions such as D-galactosamine/LPS induced mortality, doxorubicin-induced cardiotoxicity, scopolamine-induced neurological injury, and ovalbumin-triggered asthma and allergies.
  • Exhibits anti-inflammatory properties.
  • Provides anti-oxidative stress response.
  • Demonstrates anti-apoptotic effects.
  • Offers anti-cardiac cytotoxic effects.
  • Reduces mitochondrial damage.
  • Enhances mitochondrial biosynthesis.
  • Mitigates D-galactosamine/LPS induced mortality.
  • Prevents doxorubicin-induced cardiotoxicity.
  • Ameliorates scopolamine-induced neurological injury.
  • Inhibits asthma and allergies.