Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Afzelin (kaempferol-3-O-rhamnoside) | 482-39-3 | MFCD17676152 | 99.6% | 432.38 g·mol^-1 | C21H20O10 | 10 MG
SDP

Catalog No. 5000296149
Encompass_Preferred

Afzelin (kaempferol-3-O-rhamnoside) is a flavonol glycoside research reagent used to study mitochondrial metabolism, PTEN signaling, autophagy, and PI3K/Akt/mTOR pathways. It is supplied as a small-quantity, high-purity solid suitable for in vitro experiments and formulation work.

  • High purity (99.6%).
  • Molecular formula C21H20O10 and molecular weight 432.38 g·mol^-1.
  • Soluble in DMSO and ethanol; supports in vivo formulation at ≥2.5 mg/mL in recommended solvent mixtures.
  • Storage: 4°C, protect from light; solutions stable at -80°C for up to 6 months.
  • Appropriate for research on mitochondrial metabolism, PTEN, and autophagy-related pathways.

Catalog No. 50-002-96149 Supplier Medchemexpress LLC Supplier No. HYN144110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Afzelin (kaempferol-3-O-rhamnoside) is a flavonol glycoside research reagent used to study mitochondrial metabolism, PTEN signaling, autophagy, and PI3K/Akt/mTOR pathways. It is supplied as a small-quantity, high-purity solid suitable for in vitro experiments and formulation work.

  • High purity (99.6%).
  • Molecular formula C21H20O10 and molecular weight 432.38 g·mol^-1.
  • Soluble in DMSO and ethanol; supports in vivo formulation at ≥2.5 mg/mL in recommended solvent mixtures.
  • Storage: 4°C, protect from light; solutions stable at -80°C for up to 6 months.
  • Appropriate for research on mitochondrial metabolism, PTEN, and autophagy-related pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.