Learn More
TARGETMOL CHEMICALS INC AG 045572 5MG
SDP
Catalog No. 502580986
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. AG 045572 is a potent GnRH receptor antagonist that inhibits the GnRH receptor in humans and rats with Ki values of 6.0 nM and 3.8 nM respectively.AG 045572 is metabolized by CYP3A and inhibits testosterone. purity: 98%
Catalog No. 50-258-0986 Supplier TARGETMOL CHEMICALS INC Supplier No. T220255MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. AG 045572 is a potent GnRH receptor antagonist that inhibits the GnRH receptor in humans and rats with Ki values of 6.0 nM and 3.8 nM respectively.AG 045572 is metabolized by CYP3A and inhibits testosterone. purity: 98%