Learn More
Selleck Chemical LLC AG-126
SDP
Catalog No. 5000002783
Encompass_Preferred
AG126 is a tyrosine kinase inhibitor which selectively inhibits the phosphorylation of ERK1 (p44) and ERK2 (p42) at 25-50 M
Catalog No. 50-000-02783 Supplier Selleck Chemical LLC Supplier No. S65965MG
AG126 is a tyrosine kinase inhibitor which selectively inhibits the phosphorylation of ERK1 (p44) and ERK2 (p42) at 25-50 M