Learn More
Enzo Life Sciences AG-126 (5mg). CAS: 118409-62-4
SDP
Supplier:  Enzo Life Sciences BMLEI2630005
Encompass_Preferred
AG-126 prevents lipopolysaccharide-induced lethal toxicity (septic shock) in mice. Administered at 1.5 mg/mouse, it reduced LPS-induced lethal toxicity from 95% to 10%. AG-126 greatly reduces LPS-induced tyrosine phosphorylation of p42MAPK in murine peritoneal macrophages at 50 µM. EGFR, Her-2/neu receptor or PDGF-R are not inhibited at 100 µM. Inhibits tyrosine kinase-dependent B cell receptor signaling. Alternative name: 3-Hydroxy-4-nitrobenzylidenemalononitrile. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (5mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-0644