Learn More
Enzo Life Sciences AG-1288 (50mg). CAS: 116313-73-6
SDP
Supplier:  Enzo Life Sciences BMLEI3350050
Encompass_Preferred
Tyrosine kinase inhibitor. It has been shown to block TNFα induced cytotoxicity and to block glucocorticoid-induced COX-2 activity in human amnion cells (IC50=21μM). It is also reported to inhibit guanylyl cyclase. Alternative name: alpha-Cyano-(3,4-dihydroxy-5-nitro)cinnamonitrile. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (35mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0721