Learn More
Medchemexpress LLC HY-101957 10mg Medchemexpress, AG 1295 CAS:71897-07-9 Purity:>98%
SDP
Catalog No. 501967934
Encompass_Preferred
Medchemexpress, HY-101957 10mg AG 1295 CAS:71897-07-9 AG 1295 is a selective platelet-derived growth factor receptor (PDGFR) tyrosine-kinase inhibitor. AG 1295 reduces neointimal formation in aortic allograft vasculopathy by inhibition of PDGFR-beta-triggered tyrosine phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7934 Supplier Medchemexpress LLC Supplier No. HY10195710MG
Medchemexpress, HY-101957 10mg AG 1295 CAS:71897-07-9 AG 1295 is a selective platelet-derived growth factor receptor (PDGFR) tyrosine-kinase inhibitor. AG 1295 reduces neointimal formation in aortic allograft vasculopathy by inhibition of PDGFR-beta-triggered tyrosine phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.