Learn More
Apexbio Technology LLC AG-1295-25MG
SDP
Catalog No. 5000570219
Encompass_Preferred
AG-1295 (CAS No 71897-07-9) is a prominent tyrosine kinase inhibitor predominantly used in cancer research Originating from studies focusing on receptor-mediated cellular pathways AG-1295 specifically targets platelet-derived growth factor receptors (PDGFR) which play a crucial role in cell proliferation and angiogenesis By inhibiting these pathways it effectively disrupts the signaling cascade vital for tumor growth In vitro AG-1295 exhibits activity in the nanomolar to low micromolar range making it a potent agent for various experimental settings Its applications span across cell models including fibroblasts and smooth muscle cells as well as animal studies aimed at understanding disease mechanisms and testing new therapeutic strategies It is also incorporated in drug screening processes to identify potential oncological treatments Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70219 Supplier Apexbio Technology LLC Supplier No. C546525
AG-1295 (CAS No 71897-07-9) is a prominent tyrosine kinase inhibitor predominantly used in cancer research Originating from studies focusing on receptor-mediated cellular pathways AG-1295 specifically targets platelet-derived growth factor receptors (PDGFR) which play a crucial role in cell proliferation and angiogenesis By inhibiting these pathways it effectively disrupts the signaling cascade vital for tumor growth In vitro AG-1295 exhibits activity in the nanomolar to low micromolar range making it a potent agent for various experimental settings Its applications span across cell models including fibroblasts and smooth muscle cells as well as animal studies aimed at understanding disease mechanisms and testing new therapeutic strategies It is also incorporated in drug screening processes to identify potential oncological treatments Experimental concentrations typically depend on the specific research design