Learn More
Enzo Life Sciences AG-1296 (25mg). CAS: 146535-11-7
SDP
Supplier:  Enzo Life Sciences BMLEI3030025
Encompass_Preferred
A potent inhibitor of PDGF receptor tyrosine kinase (IC50=1 µM). Also inhibits FGF receptor tyrosine kinase and c-kit. Induces apoptosis in a small-cell lung cancer cell line (H526). Alternative name: Tyrphostin AG 1296, 6,7-Dimethoxy-3-phenylquinoxaline. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or dichloromethane. Long Term Storage: -20°C.
Catalog No. 50-201-0688