Learn More
Apexbio Technology LLC AG-1478 153436-53-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061598
Encompass_Preferred
AG-1478 is a selective inhibitor of epidermal growth factor receptor (EGFR) a receptor tyrosine kinase involved in cellular proliferation and survival It inhibits EGFR-mediated Erk1/2 phosphorylation induced by EGF stimulation with an IC50 of approximately 10 M in Ishikawa ovarian cell lines In experimental cancer research AG-1478 has been applied to study EGFR signaling mechanisms by reducing receptor phosphorylation and downstream pathway activation Studies using diverse cancer cell lines including ovarian (CAOV-3 SKOV-3) colorectal (SW480) hepatocellular (HA22T/VGH) and lung (A549) carcinomas employ AG-1478 to investigate EGFR s role in tumor growth and chemoresistance Additionally AG-1478 is utilized in preclinical xenograft models to assess tumor growth modulation via EGFR inhibition
Catalog No. 50-000-61598 Supplier Apexbio Technology LLC Supplier No. A83575.1
 Restricted Item
Add to Cart
Add to Cart
AG-1478 is a selective inhibitor of epidermal growth factor receptor (EGFR) a receptor tyrosine kinase involved in cellular proliferation and survival It inhibits EGFR-mediated Erk1/2 phosphorylation induced by EGF stimulation with an IC50 of approximately 10 M in Ishikawa ovarian cell lines In experimental cancer research AG-1478 has been applied to study EGFR signaling mechanisms by reducing receptor phosphorylation and downstream pathway activation Studies using diverse cancer cell lines including ovarian (CAOV-3 SKOV-3) colorectal (SW480) hepatocellular (HA22T/VGH) and lung (A549) carcinomas employ AG-1478 to investigate EGFR s role in tumor growth and chemoresistance Additionally AG-1478 is utilized in preclinical xenograft models to assess tumor growth modulation via EGFR inhibition