Learn More
TARGETMOL CHEMICALS INC AG-1478 25MG
SDP
Catalog No. 502877440
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. AG-1478 (NSC-693255) (Tyrphostin AG-1478) is a selective EGFR inhibitor. Purity 99.03%
Catalog No. 50-287-7440 Supplier TARGETMOL CHEMICALS INC Supplier No. T204725MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. AG-1478 (NSC-693255) (Tyrphostin AG-1478) is a selective EGFR inhibitor. Purity 99.03%