Learn More
Selleck Chemical LLC AG-18
SDP
Catalog No. 5000002839
Encompass_Preferred
AG-18 (RG-50810 Tyrphostin A23 TX 825) inhibits EGFR with IC50 of 35 M
Catalog No. 50-000-02839 Supplier Selleck Chemical LLC Supplier No. S800910MG
AG-18 (RG-50810 Tyrphostin A23 TX 825) inhibits EGFR with IC50 of 35 M