Learn More
Medchemexpress LLC AG-205 | 00-00-0 | 99.0% | 454.98 | C22H23ClN6OS | 5 MG
SDP
Catalog No. 5000264365
Encompass_Preferred
AG-205 is a small-molecule inhibitor of progesterone receptor membrane component-1 (PGRMC1) provided for research use. It displays anti-mitotic, anti-migratory, and anti-invasive activity, modulates apoptosis and cell-cycle regulation, and influences expression of genes involved in cholesterol biosynthesis and steroidogenesis.
  • PGRMC1 inhibitor activity relevant for cell-signaling studies.
  • Anti-mitotic, anti-migratory, and anti-invasive effects in cancer cell models.
  • Modulates apoptosis and cell-cycle regulatory pathways.
  • Alters expression of genes involved in cholesterol biosynthesis and steroidogenesis.
  • High purity (99%) suitable for research applications.
  • Stable as a powder under recommended cold storage conditions.
Catalog No. 50-002-64365 Supplier Medchemexpress LLC Supplier No. HY1156255MG
AG-205 is a small-molecule inhibitor of progesterone receptor membrane component-1 (PGRMC1) provided for research use. It displays anti-mitotic, anti-migratory, and anti-invasive activity, modulates apoptosis and cell-cycle regulation, and influences expression of genes involved in cholesterol biosynthesis and steroidogenesis.
  • PGRMC1 inhibitor activity relevant for cell-signaling studies.
  • Anti-mitotic, anti-migratory, and anti-invasive effects in cancer cell models.
  • Modulates apoptosis and cell-cycle regulatory pathways.
  • Alters expression of genes involved in cholesterol biosynthesis and steroidogenesis.
  • High purity (99%) suitable for research applications.
  • Stable as a powder under recommended cold storage conditions.