Learn More
Medchemexpress LLC HY-138630 10mg , AG-270 CAS:2201056-66-6 Purity:>98%
SDP
Catalog No. 502259508
Encompass_Preferred
HY-138630 10mg AG-270 CAS: 2201056-66-6 AG-270 is an allosteric, noncompetitive, first-in-class, reversible and orally active MAT2A inhibitor, with an IC50 of 14 nM.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9508 Supplier Medchemexpress LLC Supplier No. HY13863010MG
HY-138630 10mg AG-270 CAS: 2201056-66-6 AG-270 is an allosteric, noncompetitive, first-in-class, reversible and orally active MAT2A inhibitor, with an IC50 of 14 nM.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.