Learn More
Selleck Chemical LLC AG-270-5mg
SDP
Catalog No. 502389773
Encompass_Preferred
AG-270 is a potent and non-competitive methionine adenosyltransferase 2A (MAT2A) inhibitor which lessens the intracellular SAM levels and MTAP-null–selective antiproliferative activity.
Catalog No. 50-238-9773 Supplier Selleck Chemical LLC Supplier No. S98725MG
AG-270 is a potent and non-competitive methionine adenosyltransferase 2A (MAT2A) inhibitor which lessens the intracellular SAM levels and MTAP-null–selective antiproliferative activity.