Learn More
TARGETMOL CHEMICALS INC AG-270 5MG
SDP
Catalog No. 502579030
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AG-270 is an allosteric and orally active inhibitor of MAT2A. purity: 98%
Catalog No. 50-257-9030 Supplier TARGETMOL CHEMICALS INC Supplier No. T90505MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AG-270 is an allosteric and orally active inhibitor of MAT2A. purity: 98%