Learn More
Enzo Life Sciences AG-370 (2mg). CAS: 134036-53-6
SDP
Supplier:  Enzo Life Sciences BMLEI2290002
Encompass_Preferred
Member of the tyrphostin family of tyrosine kinase inhibitors. Selective inhibitor of the PDGF receptor kinase (IC50=20µM) vs. the EGF receptor kinase (IC50=820µM). Inhibits PDGF-induced mitogenesis in human bone marrow fibroblasts. Alternative name: Tyrphostin AG 370, Tyrphostin B7, 2-Amino-4-(1H-indol-5-yl)-1,1,3-tricyanobuta-1,3-diene. Purity: ≥99% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (2mg/ml, warm). Long Term Storage: -20°C.
Catalog No. 50-201-0621