Learn More
Selleck Chemical LLC AG-490 S1143-50mg
SDP
Catalog No. 503108211
Encompass_Preferred
AG-490 is an inhibitor of EGFR with IC50 of 0 1 M in cell-free assays 135-fold more selective for EGFR versus ErbB2 also inhibits JAK2 with no activity to Lck Lyn Btk Syk and Src
Catalog No. 50-310-8211 Supplier Selleck Chemical LLC Supplier No. S114350MG
AG-490 is an inhibitor of EGFR with IC50 of 0 1 M in cell-free assays 135-fold more selective for EGFR versus ErbB2 also inhibits JAK2 with no activity to Lck Lyn Btk Syk and Src