Learn More
Enzo Life Sciences AG-490 (50mg). CAS: 133550-30-8
SDP
Supplier:  Enzo Life Sciences BMLEI2720050
Encompass_Preferred
AG-490 is a potent inhibitor of the JAK-2 tyrosine kinase. In acute lymphoblastic leukemia (ALL) cells, which abundantly express JAK-2, AG-490 dose-dependently inhibited DNA synthesis, blocked cell growth and induced apoptosis. At 5 µM, AG-490 almost completely blocked the growth of all pre-B ALL cells but had no significant effect on the growth of mitogen-stimulated normal B or T cells, B-cell lymphoma or T-cell leukemia cells. AG-490 does not significantly inhibit other kinases such as Lck, Lyn, Btk, Syk and Src. It blocks interleukin-7-induced JAK kinase activity in T-cells (JAK-1, JAK-3) and the consequent phosphorylation of PI-3 kinase. AG-490 is cell permeable and is a valuable tool for studying the cellular role of JAK kinases in signal transduction. Alternative name: N-Benzyl-3,4-dihydroxybenzylidenecyanoacetamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (30mg/ml), dimethyl formamide (40mg/ml) or 100% ethanol (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0652