Learn More
Selleck Chemical LLC AG-490 (Tyrphostin B42) 10mg 133550-30-8 Zinc02557947
SDP
Supplier:  Selleck Chemical LLC S114310MG
Encompass_Preferred
AG-490 (Tyrphostin B42, Zinc02557947) is an inhibitor of EGFR with IC50 of 0.1 ?M in cell-free assays, 135-fold more selective for EGFR versus ErbB2, also inhibits JAK2 with no activity to Lck, Lyn, Btk, Syk and Src. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-744-6