Learn More
Abcam AG 490, JAK2/3 inhibitor, 50MG
Catalog No. 502707096
Encompass_Preferred
MW 294.3 Da, Purity >99%. JAK2/3 inhibitor (IC₅₀ values are ~10 and 25 μM, respectively). Inhibits downstream activation of STAT5a/b. Effects synaptic plasticity and cytokine-independent cell growth in vitro and spatial learning, memory and tumor cell invasion in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7096 Supplier Abcam Supplier No. AB12095050MG
MW 294.3 Da, Purity >99%. JAK2/3 inhibitor (IC₅₀ values are ~10 and 25 μM, respectively). Inhibits downstream activation of STAT5a/b. Effects synaptic plasticity and cytokine-independent cell growth in vitro and spatial learning, memory and tumor cell invasion in vivo.
The product is subject to the following: Abcam Restricted Use Statement