Learn More
Selleck Chemical LLC AG 494
SDP
Catalog No. 5000004816
Encompass_Preferred
AG-494 a member of the tyrphostin family of tyrosine kinase inhibitors is a potent inhibitor of EGF receptor autophosphorylation (IC50 1 2 M) and EGF-dependent cell growth (IC50 6 M)
Catalog No. 50-000-04816 Supplier Selleck Chemical LLC Supplier No. S65095MG
AG-494 a member of the tyrphostin family of tyrosine kinase inhibitors is a potent inhibitor of EGF receptor autophosphorylation (IC50 1 2 M) and EGF-dependent cell growth (IC50 6 M)