Learn More
Medchemexpress LLC Ag 538 | 133550-18-2 | MFCD00080276 | 99.8% | 297.26 | C16H11NO5 | 5 MG
SDP
Catalog No. 5000223348
Encompass_Preferred
AG 538 is a small-molecule, competitive inhibitor of the insulin-like growth factor-1 receptor (IGF-1R) kinase with a reported IC50 of approximately 400 nM. It is supplied as a high-purity research reagent intended for biochemical and cell-based studies.
  • Potent, competitive IGF-1R kinase inhibitor with IC50 ≈ 400 nM.
  • High purity suitable for research applications (99.8%).
  • Applicable to biochemical kinase assays and cell-based signaling studies.
  • Supplied in small, precise sample sizes for laboratory testing.
  • Molecular formula C16H11NO5; molecular weight 297.26.
Catalog No. 50-002-23348 Supplier Medchemexpress LLC Supplier No. HY1354365MG
AG 538 is a small-molecule, competitive inhibitor of the insulin-like growth factor-1 receptor (IGF-1R) kinase with a reported IC50 of approximately 400 nM. It is supplied as a high-purity research reagent intended for biochemical and cell-based studies.
  • Potent, competitive IGF-1R kinase inhibitor with IC50 ≈ 400 nM.
  • High purity suitable for research applications (99.8%).
  • Applicable to biochemical kinase assays and cell-based signaling studies.
  • Supplied in small, precise sample sizes for laboratory testing.
  • Molecular formula C16H11NO5; molecular weight 297.26.