Learn More
Medchemexpress LLC HY-15336 100mg Medchemexpress, AG 555 CAS:133550-34-2 Purity:>98%
SDP
Catalog No. 502081707
Encompass_Preferred
Medchemexpress, HY-15336 100mg AG 555 CAS:133550-34-2 AG 555 (Tyrphostin AG 555), a potent antiretroviral drug, is a potent and selective inhibitor of EGFR and blocks Cdk2 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1707 Supplier Medchemexpress LLC Supplier No. HY15336100MG
Medchemexpress, HY-15336 100mg AG 555 CAS:133550-34-2 AG 555 (Tyrphostin AG 555), a potent antiretroviral drug, is a potent and selective inhibitor of EGFR and blocks Cdk2 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.