Learn More
Selleck Chemical LLC AG 555
SDP
Catalog No. 5000002808
Encompass_Preferred
AG-555 (Tyrphostin B46) is an inhibitor of tyrosine kinase interacts directly with topoisomerase I thus preventing DNA relaxation It inhibits EGFR with an IC50 of 0 7 M
Catalog No. 50-000-02808 Supplier Selleck Chemical LLC Supplier No. S65255MG
AG-555 (Tyrphostin B46) is an inhibitor of tyrosine kinase interacts directly with topoisomerase I thus preventing DNA relaxation It inhibits EGFR with an IC50 of 0 7 M