Learn More
Medchemexpress LLC AG 555 (Tyrphostin AG 555) | 133550-34-2 | C19H18N2O3 | 1 ML
SDP
Catalog No. 5000372868
Encompass_Preferred
AG 555 (Tyrphostin AG 555) is a potent antiretroviral agent that acts as a selective inhibitor of EGFR and blocks Cdk2 activation. This product is for research use only.
  • Potent antiretroviral agent
  • Selective inhibitor of EGFR
  • Blocks Cdk2 activation
  • Induces growth arrest at G1-S and early S
  • Effective in arresting growth of EGFR overexpressor cells
  • Suppresses BPV-1 transcription
  • Inhibits Moloney murine leukemia virus replication
Catalog No. 50-003-72868 Supplier Medchemexpress LLC Supplier No. HY1533610MM/1ML
AG 555 (Tyrphostin AG 555) is a potent antiretroviral agent that acts as a selective inhibitor of EGFR and blocks Cdk2 activation. This product is for research use only.
  • Potent antiretroviral agent
  • Selective inhibitor of EGFR
  • Blocks Cdk2 activation
  • Induces growth arrest at G1-S and early S
  • Effective in arresting growth of EGFR overexpressor cells
  • Suppresses BPV-1 transcription
  • Inhibits Moloney murine leukemia virus replication