Learn More
Medchemexpress LLC AG-636 | 1623416-31-8 | 98.07% | 343.38 | 1 ML
SDP
Catalog No. 5000350997
Encompass_Preferred
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor. With an IC50 of 17 nM, it demonstrates strong anticancer effects and growth-inhibitory activity in various cancer cell lines. It has also shown robust tumor growth inhibition in xenograft lymphoma models.
  • Potent, reversible, selective, and orally active DHODH inhibitor
  • IC50 of 17 nM against DHODH
  • Strong growth-inhibitory activity in cancer cell lines of hematologic versus solid tumor origin
  • Robust tumor growth inhibition in xenograft lymphoma models
  • Currently in phase 1 clinical trial for lymphoma
Catalog No. 50-003-50997 Supplier Medchemexpress LLC Supplier No. HY13746310MM/1ML
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor. With an IC50 of 17 nM, it demonstrates strong anticancer effects and growth-inhibitory activity in various cancer cell lines. It has also shown robust tumor growth inhibition in xenograft lymphoma models.
  • Potent, reversible, selective, and orally active DHODH inhibitor
  • IC50 of 17 nM against DHODH
  • Strong growth-inhibitory activity in cancer cell lines of hematologic versus solid tumor origin
  • Robust tumor growth inhibition in xenograft lymphoma models
  • Currently in phase 1 clinical trial for lymphoma