Learn More
Medchemexpress LLC AG-636 | 1623416-31-8 | 98.1% | 343.38 | 100 MG
SDP
Catalog No. 5000350169
Encompass_Preferred
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor. It demonstrates strong anticancer effects by exhibiting robust growth-inhibitory activity in cancer cell lines and significant tumor growth inhibition in xenograft models.
  • Potent, reversible, selective, and orally active DHODH inhibitor with an IC50 of 17 nM.
  • Exhibits strong anticancer effects.
  • Demonstrates robust growth-inhibitory activity in cancer cell lines.
  • Leads to significant tumor growth inhibition in DLBCL tumor xenograft model.
Catalog No. 50-003-50169 Supplier Medchemexpress LLC Supplier No. HY137463100MG
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor. It demonstrates strong anticancer effects by exhibiting robust growth-inhibitory activity in cancer cell lines and significant tumor growth inhibition in xenograft models.
  • Potent, reversible, selective, and orally active DHODH inhibitor with an IC50 of 17 nM.
  • Exhibits strong anticancer effects.
  • Demonstrates robust growth-inhibitory activity in cancer cell lines.
  • Leads to significant tumor growth inhibition in DLBCL tumor xenograft model.