Learn More
Medchemexpress LLC AG-636 | 1623416-31-8 | 98.1% | 343.38 | 50 MG
SDP
Catalog No. 5000352393
Encompass_Preferred
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor with an IC50 of 17 nM. It demonstrates strong anticancer effects.
  • Exhibits significant growth-inhibitory activity in cancer cell lines.
  • Results in robust tumor growth inhibition in xenograft lymphoma tumor models.
  • Suitable for laboratory research applications.
Catalog No. 50-003-52393 Supplier Medchemexpress LLC Supplier No. HY13746350MG
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor with an IC50 of 17 nM. It demonstrates strong anticancer effects.
  • Exhibits significant growth-inhibitory activity in cancer cell lines.
  • Results in robust tumor growth inhibition in xenograft lymphoma tumor models.
  • Suitable for laboratory research applications.