Learn More
TARGETMOL CHEMICALS INC AG-636 5MG
SDP
Catalog No. 502878075
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AG-636 is a potent, reversible, and selective DHODH inhibitor with an IC50 of 17 nM and is orally active. It demonstrates strong anticancer effects. Purity 99.19%
Catalog No. 50-287-8075 Supplier TARGETMOL CHEMICALS INC Supplier No. T91215MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AG-636 is a potent, reversible, and selective DHODH inhibitor with an IC50 of 17 nM and is orally active. It demonstrates strong anticancer effects. Purity 99.19%