Learn More
World Precision Instrument Ag/AgCl Electrode-EP05
SDP
Catalog No. 50821858
Encompass
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
Catalog No. 50-821-858 Supplier World Precision Instrument Supplier No. EP05
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.