Learn More
World Precision Instrument Ag/AgCl Electrode-EP12
SDP
Catalog No. 50821861
Encompass
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
Catalog No. 50-821-861 Supplier World Precision Instrument Supplier No. EP12
 Restricted Item
Add to Cart
Add to Cart
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.