Learn More
World Precision Instrument Ag/AgCl Electrode-EP1
SDP
Catalog No. 50821860
Encompass
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
Catalog No. 50-821-860 Supplier World Precision Instrument Supplier No. EP1
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.