Learn More
Perkin Elmer US LLC Silver (Ag) - Chromium (Cr) - Copper (Cu) - Nickel (Ni) Lumina Hollow Cathode Lamp
Supplier:  Perkin Elmer US LLC N3050210
Encompass_Preferred
Multi-Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Silver (Ag), Chromium (Cr), Copper (Cu) and Nickel (Ni). Please note that multi-element lamps are not designed to be used in a furnace system. Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.
Catalog No. 50-904-4282