Learn More
Strem, An Ascensus Company CAS# [165461-74-5]. 1g. Triethylphosphine(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)silver(I), min. 98%. MFCD11973810
SDP
Catalog No. 501979367
Encompass
CAS# [165461-74-5]. 1g. Triethylphosphine(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)silver(I), min. 98%. MFCD11973810. Molecular Weight: 521.20. Molecular Formula: Ag(C3F7COCHCOC4H9)P(CH2CH3)3. Color/form: yellow liq. to low melting solid Strem# 47-3025. www.strem.com/catalog/v3/47-3025/
Catalog No. 50-197-9367 Supplier Strem, An Ascensus Company Supplier No. 4730251G
Add to Cart
Add to Cart
CAS# [165461-74-5]. 1g. Triethylphosphine(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)silver(I), min. 98%. MFCD11973810. Molecular Weight: 521.20. Molecular Formula: Ag(C3F7COCHCOC4H9)P(CH2CH3)3. Color/form: yellow liq. to low melting solid Strem# 47-3025. www.strem.com/catalog/v3/47-3025/